missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ S41i Incubator Shaker Cell Line Bundle (EU)

The New Brunswick S41i is a CO2 incubator with an integrated shaker. An additional perforated shelf that can be used for static incubation at the same time as shaking. Contamination protection is supported by a high temperature disinfection, the fanless design and the seamless chamber.

Marca:  Eppendorf™ S41I230111


Dettagli aggiuntivi : Peso : 165.00000kg

Codice prodotto. 17382970

  • 21025.00€ / Pezzo
Spedizione stimata: 22-08-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

  • mAb-Discovery Bundle: Cell Culture Versatility Solution, for Hybridoma cultures, antibody expression, and cell line establishment
  • Bundle consists of: 1x New Brunswick™ S41i, 170 L, (25 mm/1 in. orbit), 1x universal platform and 2x sticky pads, 230 V, 50 Hz
  • Experimental flexibility: Perforated shelf allows for static incubation at the same time as shaking
  • Stable, uniform and vibration-free shaking: robust Innova triple eccentric drive
  • Saves gas and provides an optimal growing atmosphere: Smart gas control (0.2 - 20% CO2)
  • Ensures surface sterility: automatic high temperature disinfection and inner/outer door
  • Fast and reliable cleaning: Smooth, seamless chamber
  • Increasable capacity: Two devices stackable
  • Precise and easy control: Intuitive touch screen
  • Humidification included
  • Shaking speed 25 - 400 rpm (25 - 250 rpm if stacked)
Specifica

Specifica

25 mm
230 V
170 L
28.7 in.
73 cm
33.5 in.
87.5 cm
Touchscreen
Continuo
25 to 400 rpm (25 to 250 rpm if stacked)
4°C to 50°C
50 Hz
17.5 in.
44.4 cm
34.4 in.
69.1 cm
Ethernet
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali