CAS RN 100-56-1
CAS RN 100-56-1
IUPAC Name:
chloro(phenyl)mercury
Sinonimi:
phenyl mercuric chloride
Peso molecolare (g/mol):
313.15
Formula molecolare:
C6H5ClHg
InChi Key:
AWGTVRDHKJQFAX-UHFFFAOYSA-M
SMILES:
Cl[Hg]C1=CC=CC=C1
1
–
3
di
3
risultati
1
Phenylmercury(II) chloride, 96%, Hg 63.5%, Thermo Scientific Chemicals
CAS: 100-56-1 Formula molecolare: C6H5ClHg Molecular Weight (g/mol): 313.15 Numero MDL: MFCD00000463 InChI Key: AWGTVRDHKJQFAX-UHFFFAOYSA-M Sinonimo: phenylmercuric chloride,agrenal,phenylmercury chloride,mercury, chlorophenyl,hexason,mercutal,merfazin,stopspot,agronal,mersolite 2 PubChem CID: 7511 IUPAC Name: chloro(phenyl)mercury SMILES: Cl[Hg]C1=CC=CC=C1
2
Buffer concentrate pH 5.00, For 500 ml buffer solution, citric acid / sodium hydroxide solution, Honeywell Fluka™
for 500 ml buffer solution, citric acid / sodium hydroxide solution