CAS RN 100-80-1
CAS RN 100-80-1
IUPAC Name:
1-ethenyl-3-methylbenzene
Sinonimi:
M-methylstyrene
Peso molecolare (g/mol):
118.18
Formula molecolare:
C9H10
InChi Key:
JZHGRUMIRATHIU-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(C=C)=C1
1
–
2
di
2
risultati
1
3-Methylstyrene, 95%, Thermo Scientific Chemicals
CAS: 100-80-1 Formula molecolare: C9H10 Molecular Weight (g/mol): 118.18 InChI Key: JZHGRUMIRATHIU-UHFFFAOYSA-N Sinonimo: 3-methylstyrene,3-vinyltoluene,1-methyl-3-vinylbenzene,m-vinyltoluene,m-methylstyrene,benzene, 1-ethenyl-3-methyl,styrene, m-methyl,m-methyl styrene,meta-methylstyrene,unii-ffk4i9430g PubChem CID: 7529 IUPAC Name: 1-ethenyl-3-methylbenzene SMILES: CC1=CC=CC(=C1)C=C