CAS RN 100-99-2
CAS RN 100-99-2
IUPAC Name:
tris(2-methylpropyl)alumane
Sinonimi:
triisobutylaluminium
Peso molecolare (g/mol):
198.33
Formula molecolare:
C12H27Al
InChi Key:
MCULRUJILOGHCJ-UHFFFAOYSA-N
SMILES:
CC(C)C[Al](CC(C)C)CC(C)C
1
–
1
di
1
risultati
1
Triisobutylaluminium, 1.1M solution in toluene, AcroSeal™, Thermo Scientific Chemicals
CAS: 100-99-2 Formula molecolare: C12H27Al Molecular Weight (g/mol): 198.33 Numero MDL: MFCD00008929 InChI Key: MCULRUJILOGHCJ-UHFFFAOYSA-N Sinonimo: triisobutylaluminum,triisobutylaluminium,aluminum, tris 2-methylpropyl,tibal,triisobutylalane,triisobutyl aluminum,aluminum, triisobutyl,tris isobutyl alane,tris isobutyl aluminum,unii-09p2thv2x4 PubChem CID: 16682931 IUPAC Name: tris(2-methylpropyl)alumane SMILES: CC(C)C[Al](CC(C)C)CC(C)C
Promozioni disponibili