CAS RN 100032-79-9
CAS RN 100032-79-9
IUPAC Name:
(4-bromo-2-nitrophenyl)hydrazine hydrochloride
Sinonimi:
(4-bromo-2-nitrophenyl)hydrazine hydrochloride
Peso molecolare (g/mol):
268.5
Formula molecolare:
C6H7BrClN3O2
InChi Key:
SIBURLORIWZQNT-UHFFFAOYSA-N
SMILES:
Cl.NNC1=C(C=C(Br)C=C1)[N+]([O-])=O
1
–
1
di
1
risultati
1
4-Bromo-2-nitrophenylhydrazine hydrochloride, 95%, Thermo Scientific Chemicals
CAS: 100032-79-9 Formula molecolare: C6H7BrClN3O2 Molecular Weight (g/mol): 268.50 Numero MDL: MFCD09260482 InChI Key: SIBURLORIWZQNT-UHFFFAOYSA-N Sinonimo: 4-bromo-2-nitrophenylhydrazine hydrochloride,4-bromo-2-nitrophenyl hydrazine hydrochloride,acmc-20amnd,4-bromo-2-nitrophenyl hydrazine;hydrochloride,4-bromo-2-nitrophenylhydrazinehydrochloride,4-bromo-2-nitrophenyl hydrazine-hydrogen chloride 1/1 PubChem CID: 44119379 IUPAC Name: (4-bromo-2-nitrophenyl)hydrazine;hydrochloride SMILES: Cl.NNC1=C(C=C(Br)C=C1)[N+]([O-])=O