CAS RN 100124-06-9
CAS RN 100124-06-9
IUPAC Name:
{8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-6-yl}boronic acid
Sinonimi:
8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-6-ylboronic acid
Peso molecolare (g/mol):
212.01
Formula molecolare:
C12H9BO3
InChi Key:
ZXHUJRZYLRVVNP-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC=CC2=C1OC1=C2C=CC=C1
1
–
1
di
1
risultati
1
Dibenzofuran-4-boronic acid, 98+%, Thermo Scientific Chemicals
CAS: 100124-06-9 Formula molecolare: C12H9BO3 Molecular Weight (g/mol): 212.011 Numero MDL: MFCD00092336 InChI Key: ZXHUJRZYLRVVNP-UHFFFAOYSA-N Sinonimo: dibenzofuran-4-boronic acid,dibenzo b,d furan-4-ylboronic acid,4-dibenzofuranyl boronic acid,4-dibenzofuranboronic acid,4-dibenzofuranboronicacid,4-dibenzofuranylboronic acid,boronic acid, 4-dibenzofuranyl,dibenzo b,d furan-4-boronic acid,8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-6-ylboronic acid PubChem CID: 2734328 IUPAC Name: dibenzofuran-4-ylboronic acid SMILES: B(C1=C2C(=CC=C1)C3=CC=CC=C3O2)(O)O