CAS RN 1001907-57-8
CAS RN 1001907-57-8
IUPAC Name:
(2-methyl-2H-indazol-6-yl)boronic acid
Sinonimi:
2-methylindazol-6-ylboronic acid
Peso molecolare (g/mol):
175.98
Formula molecolare:
C8H9BN2O2
InChi Key:
JADBVQWCMHPPQA-UHFFFAOYSA-N
SMILES:
CN1C=C2C=CC(=CC2=N1)B(O)O