CAS RN 10026-18-3
CAS RN 10026-18-3
IUPAC Name:
cobalt(3+) trifluoride
Sinonimi:
cobalt(III) fluoride
Peso molecolare (g/mol):
115.93
Formula molecolare:
CoF3
InChi Key:
WZJQNLGQTOCWDS-UHFFFAOYSA-K
SMILES:
[F-].[F-].[F-].[Co+3]
1
–
1
di
1
risultati
1
Cobalt(III) fluoride, 99%, Thermo Scientific Chemicals
CAS: 10026-18-3 Formula molecolare: CoF3 Numero MDL: MFCD00010942