CAS RN 1003043-34-2
CAS RN 1003043-34-2
IUPAC Name:
(6-bromo-5-methylpyridin-3-yl)boronic acid
Sinonimi:
6-bromo-5-methylpyridin-3-ylboronic acid
Peso molecolare (g/mol):
215.84
Formula molecolare:
C6H7BBrNO2
InChi Key:
IPCXBKSYMWZLBR-UHFFFAOYSA-N
SMILES:
CC1=C(Br)N=CC(=C1)B(O)O
1
–
2
di
2
risultati
2
6-Bromo-5-methylpyridine-3-boronic acid, 95%, Thermo Scientific™
CAS: 1003043-34-2 Formula molecolare: C6H7BBrNO2 Molecular Weight (g/mol): 215.841 Numero MDL: MFCD08274468 InChI Key: IPCXBKSYMWZLBR-UHFFFAOYSA-N Sinonimo: 2-bromo-3-methylpyridine-5-boronic acid,6-bromo-5-methylpyridine-3-boronic acid,6-bromo-5-methylpyridin-3-yl boronic acid,2-bromo-3-methyl-5-pyridineboronic acid,5-borono-2-bromo-3-picoline,6-bromo-5-methylpyridin-3-yl-3-boronic acid,2-bromo-3-picoline-5-boronic acid,6-bromo-5-methyl-3-pyridyl boronic acid,pubchem5081,acmc-2097oc PubChem CID: 17750219 IUPAC Name: (6-bromo-5-methylpyridin-3-yl)boronic acid SMILES: B(C1=CC(=C(N=C1)Br)C)(O)O