CAS RN 10031-26-2
CAS RN 10031-26-2
IUPAC Name:
iron(3+) tribromide
Sinonimi:
iron(III) bromide
Peso molecolare (g/mol):
295.56
Formula molecolare:
Br3Fe
InChi Key:
FEONEKOZSGPOFN-UHFFFAOYSA-K
SMILES:
[Fe+3].[Br-].[Br-].[Br-]
1
–
3
di
3
risultati
1
Iron(III) bromide, anhydrous, 98+%, Thermo Scientific Chemicals
CAS: 10031-26-2 Numero MDL: MFCD00016082
3
Iron(III) bromide, 90+%, Thermo Scientific Chemicals
CAS: 10031-26-2 Formula molecolare: Br3Fe Molecular Weight (g/mol): 295.56 Numero MDL: MFCD00016082