CAS RN 10042-76-9
CAS RN 10042-76-9
Strontium nitrate, 99+%, for analysis, Thermo Scientific Chemicals
CAS: 10042-76-9 Formula molecolare: N2O6Sr Molecular Weight (g/mol): 211.63 Numero MDL: MFCD00011248 InChI Key: DHEQXMRUPNDRPG-UHFFFAOYSA-N Sinonimo: strontium nitrate,strontium dinitrate,nitric acid, strontium salt,strontium ii nitrate 1:2,unii-bdg873aqzl,nitrate de strontium french,hsdb 787,strontium nitrate sr no3 2,bdg873aqzl,nitric acid, strontium salt 2:1 PubChem CID: 24848 SMILES: [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Promozioni disponibili
Strontium nitrate, 98%, extra pure, Thermo Scientific Chemicals
CAS: 10042-76-9 Formula molecolare: N2O6Sr Molecular Weight (g/mol): 211.63 Numero MDL: MFCD00011248 InChI Key: DHEQXMRUPNDRPG-UHFFFAOYSA-N Sinonimo: strontium nitrate,strontium dinitrate,nitric acid, strontium salt,strontium ii nitrate 1:2,unii-bdg873aqzl,nitrate de strontium french,hsdb 787,strontium nitrate sr no3 2,bdg873aqzl,nitric acid, strontium salt 2:1 PubChem CID: 24848 IUPAC Name: strontium(2+) dinitrate SMILES: [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Strontium nitrate, anhydrous, 98%, Thermo Scientific Chemicals
CAS: 10042-76-9 Formula molecolare: N2O6Sr Molecular Weight (g/mol): 211.63 Numero MDL: MFCD00011248 InChI Key: DHEQXMRUPNDRPG-UHFFFAOYSA-N Sinonimo: strontium nitrate,strontium dinitrate,nitric acid, strontium salt,strontium ii nitrate 1:2,unii-bdg873aqzl,nitrate de strontium french,hsdb 787,strontium nitrate sr no3 2,bdg873aqzl,nitric acid, strontium salt 2:1 PubChem CID: 24848 IUPAC Name: strontium;dinitrate SMILES: [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Strontium nitrate, Puratronic™, 99.9965% (metals basis), Thermo Scientific Chemicals
CAS: 10042-76-9 Formula molecolare: N2O6Sr Molecular Weight (g/mol): 211.63 Numero MDL: MFCD00011248 InChI Key: DHEQXMRUPNDRPG-UHFFFAOYSA-N Sinonimo: strontium nitrate,strontium dinitrate,nitric acid, strontium salt,strontium ii nitrate 1:2,unii-bdg873aqzl,nitrate de strontium french,hsdb 787,strontium nitrate sr no3 2,bdg873aqzl,nitric acid, strontium salt 2:1 PubChem CID: 24848 SMILES: [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Strontium nitrate, 99+%, ACS reagent, Thermo Scientific Chemicals
CAS: 10042-76-9 Formula molecolare: N2O6Sr Molecular Weight (g/mol): 211.63 Numero MDL: MFCD00011248 InChI Key: DHEQXMRUPNDRPG-UHFFFAOYSA-N Sinonimo: strontium nitrate,strontium dinitrate,nitric acid, strontium salt,strontium ii nitrate 1:2,unii-bdg873aqzl,nitrate de strontium french,hsdb 787,strontium nitrate sr no3 2,bdg873aqzl,nitric acid, strontium salt 2:1 PubChem CID: 24848 SMILES: [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Strontium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 10042-76-9 Formula molecolare: N2O6Sr Molecular Weight (g/mol): 211.63 Numero MDL: MFCD00011248 InChI Key: DHEQXMRUPNDRPG-UHFFFAOYSA-N Sinonimo: strontium nitrate,strontium dinitrate,nitric acid, strontium salt,strontium ii nitrate 1:2,unii-bdg873aqzl,nitrate de strontium french,hsdb 787,strontium nitrate sr no3 2,bdg873aqzl,nitric acid, strontium salt 2:1 PubChem CID: 24848 IUPAC Name: strontium(2+) dinitrate SMILES: [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Strontium nitrate, ACS, 99.0% min, Thermo Scientific Chemicals
CAS: 10042-76-9 Formula molecolare: N2O6Sr Molecular Weight (g/mol): 211.63 Numero MDL: MFCD00011248 InChI Key: DHEQXMRUPNDRPG-UHFFFAOYSA-N Sinonimo: strontium nitrate,strontium dinitrate,nitric acid, strontium salt,strontium ii nitrate 1:2,unii-bdg873aqzl,nitrate de strontium french,hsdb 787,strontium nitrate sr no3 2,bdg873aqzl,nitric acid, strontium salt 2:1 PubChem CID: 24848 SMILES: [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Strontium nitrate, anhydrous, 99.97% (metals basis), Thermo Scientific Chemicals
CAS: 10042-76-9 Formula molecolare: N2O6Sr Molecular Weight (g/mol): 211.63 Numero MDL: MFCD00011248 InChI Key: DHEQXMRUPNDRPG-UHFFFAOYSA-N Sinonimo: strontium nitrate,strontium dinitrate,nitric acid, strontium salt,strontium ii nitrate 1:2,unii-bdg873aqzl,nitrate de strontium french,hsdb 787,strontium nitrate sr no3 2,bdg873aqzl,nitric acid, strontium salt 2:1 PubChem CID: 24848 SMILES: [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Strontium nitrate, ACS Reagent, ≥99.0%, Honeywell Fluka™
CAS: 10042-76-9 Formula molecolare: N2O6Sr Molecular Weight (g/mol): 211.63 Numero MDL: MFCD00011248 InChI Key: DHEQXMRUPNDRPG-UHFFFAOYSA-N Sinonimo: strontium nitrate,strontium dinitrate,nitric acid, strontium salt,strontium ii nitrate 1:2,unii-bdg873aqzl,nitrate de strontium french,hsdb 787,strontium nitrate sr no3 2,bdg873aqzl,nitric acid, strontium salt 2:1 PubChem CID: 24848 SMILES: [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Strontium nitrate, Puriss., ≥98%, Honeywell Fluka™
CAS: 10042-76-9 Formula molecolare: N2O6Sr Molecular Weight (g/mol): 211.63 Numero MDL: MFCD00011248 InChI Key: DHEQXMRUPNDRPG-UHFFFAOYSA-N Sinonimo: strontium nitrate,strontium dinitrate,nitric acid, strontium salt,strontium ii nitrate 1:2,unii-bdg873aqzl,nitrate de strontium french,hsdb 787,strontium nitrate sr no3 2,bdg873aqzl,nitric acid, strontium salt 2:1 PubChem CID: 24848 SMILES: [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O