CAS RN 10051-44-2
CAS RN 10051-44-2
IUPAC Name:
sodium hexanoate
Sinonimi:
sodium hexanoate
Peso molecolare (g/mol):
138.14
Formula molecolare:
C6H11NaO2
InChi Key:
UDWXLZLRRVQONG-UHFFFAOYSA-M
SMILES:
[Na+].CCCCCC([O-])=O
1
–
1
di
1
risultati
1
Sodium hexanoate, 99%, Thermo Scientific Chemicals
CAS: 10051-44-2 Formula molecolare: C6H11NaO2 Molecular Weight (g/mol): 138.14 Numero MDL: MFCD00059056 InChI Key: UDWXLZLRRVQONG-UHFFFAOYSA-M Sinonimo: sodium hexanoate,sodium caproate,hexanoic acid, sodium salt,hexanoic acid sodium salt,sodium capronate,sodium n-caproate,sodium n-hexanoate,unii-e00cg11s66,caproic acid sodium salt,hexanoic acid, sodium salt 1:1 PubChem CID: 4087444 IUPAC Name: sodium;hexanoate SMILES: [Na+].CCCCCC([O-])=O