CAS RN 100986-89-8
CAS RN 100986-89-8
IUPAC Name:
6,7-difluoro-2-methyl-10-oxo-4-oxa-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5(13),6,8,11-tetraene-11-carboxylic acid
Sinonimi:
6,7-difluoro-2-methyl-10-oxo-4-oxa-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5(13),6,8,11-tetraene-11-carboxylic acid
Peso molecolare (g/mol):
281.22
Formula molecolare:
C13H9F2NO4
InChi Key:
NVKWWNNJFKZNJO-UHFFFAOYNA-N
SMILES:
CC1COC2=C3N1C=C(C(O)=O)C(=O)C3=CC(F)=C2F
1
–
1
di
1
risultati
1
(S)-(-)-9,10-Difluoro-3-methyl-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, 97+%, Thermo Scientific™
CAS: 100986-89-8 Formula molecolare: C13H9F2NO4 Molecular Weight (g/mol): 281.215 Numero MDL: MFCD04039905 InChI Key: NVKWWNNJFKZNJO-YFKPBYRVSA-N Sinonimo: levofloxacin q-acid,levofloxacin carboxylic acid,s-9,10-difluoro-3-methyl-7-oxo-3,7-dihydro-2h-1,4 oxazino 2,3,4-ij quinoline-6-carboxylic acid,unii-08gt8fy84e,levofloxacin related compound b,levofloxacin acid,3s-9,10-difluoro-2,3-dihydro-3-methyl-7-oxo-7h-pyrido 1,2,3-de-1,4-benzoxazine-6-carboxylic acid,levofloxacin q-acid,-,s-9,10-difluoro-3-methyl-7-oxo-2,3-dihydro-7h-pyrido 1,2,3-de-1,4-benzoxazine-6-carboxylic acid,levofloxacin related compound b usp PubChem CID: 688333 SMILES: CC1COC2=C3N1C=C(C(=O)C3=CC(=C2F)F)C(=O)O