CAS RN 10102-45-1
CAS RN 10102-45-1
IUPAC Name:
λ¹-thallanylium nitrate
Sinonimi:
λ¹-thallanylium nitrate
Peso molecolare (g/mol):
266.38
Formula molecolare:
NO3Tl
InChi Key:
FYWSTUCDSVYLPV-UHFFFAOYSA-N
SMILES:
[Tl+].[O-][N+]([O-])=O
1
–
4
di
4
risultati
1
Thallium(I) nitrate, 99.5%, Thermo Scientific Chemicals
CAS: 10102-45-1 Formula molecolare: NO3Tl Molecular Weight (g/mol): 266.38 Numero MDL: MFCD00011280 InChI Key: FYWSTUCDSVYLPV-UHFFFAOYSA-N Sinonimo: thallium i nitrate,thallium 1+ nitrate,thallous nitrate,thallium mononitrate,thallium nitrate van,thallium nitrate, tlno3,unii-ljq38dsr12,rcra waste number u217,thallium i nitrate 1:1 PubChem CID: 24937 IUPAC Name: thallium(1+);nitrate SMILES: [Tl+].[O-][N+]([O-])=O