CAS RN 101084-96-2
CAS RN 101084-96-2
IUPAC Name:
2-methoxy-4-nitrobenzonitrile
Sinonimi:
2-methoxy-4-nitrobenzonitrile
Peso molecolare (g/mol):
178.15
Formula molecolare:
C8H6N2O3
InChi Key:
MLIKCKXLGYEGAO-UHFFFAOYSA-N
SMILES:
COC1=C(C=CC(=C1)[N+]([O-])=O)C#N
1
–
2
di
2
risultati
2
2-Methoxy-4-nitrobenzonitrile, 98%, Thermo Scientific Chemicals
CAS: 101084-96-2 Formula molecolare: C8H6N2O3 Molecular Weight (g/mol): 178.147 Numero MDL: MFCD02093781 InChI Key: MLIKCKXLGYEGAO-UHFFFAOYSA-N Sinonimo: 2-cyano-5-nitroanisole,acmc-20a4y1,4-nitro-2-methoxybenzonitrile,2-methoxy-4-nitro-benzonitrile,2-methyloxy-4-nitrobenzonitrile,2-methoxy-4-nitrobenzonitrile PubChem CID: 291867 IUPAC Name: 2-methoxy-4-nitrobenzonitrile SMILES: COC1=C(C=CC(=C1)[N+](=O)[O-])C#N