CAS RN 101316-46-5
CAS RN 101316-46-5
IUPAC Name:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
11
di
11
risultati
1
Petroleum Ether, puriss. p.a. ACS Reagent, Honeywell™
CAS: 101316-46-5 Numero MDL: MFCD00081849
2
Petroleum Ether, puriss. p.a., Reag. Ph. Eur., Honeywell™
CAS: 101316-46-5 Numero MDL: MFCD00081849
3
Petroleum Ether, puriss. p.a., Honeywell™
CAS: 101316-46-5 Numero MDL: MFCD00081849
4
Petroleum ether, For pesticide residue analysis, low boiling point hydrogen treated naphtha, bp 40-60 °C, Honeywell Riedel-de Haën™
CAS: 101316-46-5 Numero MDL: MFCD00081849
5
Petroleum Ether, puriss., Meets analytical specification of DAB, Honeywell™
CAS: 101316-46-5 Numero MDL: MFCD00081849
7
Petroleum ether, low boiling point, Honeywell™
CAS: 101316-46-5 Numero MDL: MFCD00081849
8
Petroleum Ether, puriss., Honeywell™ Riedel-de Haën™
CAS: 101316-46-5 Numero MDL: MFCD00081849
9
Petroleum Ether, puriss. p.a., ACS Reagent, Reag. ISO, Honeywell™ Riedel-de Haën™
CAS: 101316-46-5 Numero MDL: MFCD00081849
10
Fam Benzine, Honeywell
CAS: 101316-46-5 Numero MDL: MFCD00081849