CAS RN 1016-05-3
CAS RN 1016-05-3
IUPAC Name:
8λ⁶-thiatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaene-8,8-dione
Sinonimi:
8λ⁶-thiatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaene-8,8-dione
Peso molecolare (g/mol):
216.25
Formula molecolare:
C12H8O2S
InChi Key:
IKJFYINYNJYDTA-UHFFFAOYSA-N
SMILES:
O=S1(=O)C2=CC=CC=C2C2=CC=CC=C12