CAS RN 101689-18-3
CAS RN 101689-18-3
IUPAC Name:
methyl 7-phenylheptanoate
Sinonimi:
methyl 7-phenylheptanoate
Peso molecolare (g/mol):
220.31
Formula molecolare:
C14H20O2
InChi Key:
FZSNATXOGRAOIL-UHFFFAOYSA-N
SMILES:
COC(=O)CCCCCCC1=CC=CC=C1
1
–
1
di
1
risultati
1
Methyl 7-phenylheptanoate, 98%, Thermo Scientific Chemicals
CAS: 101689-18-3 Formula molecolare: C14H20O2 Molecular Weight (g/mol): 220.31 Numero MDL: MFCD02093971 InChI Key: FZSNATXOGRAOIL-UHFFFAOYSA-N Sinonimo: acmc-20akft,benzeneheptanoic acid,methyl ester,7-phenylheptanoic acid methyl ester PubChem CID: 7010318 IUPAC Name: methyl 7-phenylheptanoate SMILES: COC(=O)CCCCCCC1=CC=CC=C1