CAS RN 101772-29-6
CAS RN 101772-29-6
IUPAC Name:
2-{[(tert-butoxy)carbonyl](methyl)amino}-3-hydroxypropanoic acid
Sinonimi:
2-[(tert-butoxycarbonyl)(methyl)amino]-3-hydroxypropanoic acid
Peso molecolare (g/mol):
219.24
Formula molecolare:
C9H17NO5
InChi Key:
NJQGIDVCNBZXLG-UHFFFAOYNA-N
SMILES:
CN(C(CO)C(O)=O)C(=O)OC(C)(C)C
1
–
1
di
1
risultati
1
N-Boc-N-methyl-L-serine, 98%, Thermo Scientific Chemicals
CAS: 101772-29-6 Formula molecolare: C9H17NO5 Molecular Weight (g/mol): 219.237 Numero MDL: MFCD00037249 InChI Key: NJQGIDVCNBZXLG-LURJTMIESA-N Sinonimo: boc-n-me-ser-oh,boc-l-meser-oh,boc-n-a-methyl-l-serine,boc-n-,boc-n-,s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid,s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid,l-serine,n-1,1-dimethylethoxy carbonyl-n-methyl,boc-meser-oh,2s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid,2s-2-tert-butoxycarbonyl methyl amino-3-hydroxypropanoic acid PubChem CID: 7009594 IUPAC Name: (2S)-3-hydroxy-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid SMILES: CC(C)(C)OC(=O)N(C)C(CO)C(=O)O