CAS RN 1017779-39-3
CAS RN 1017779-39-3
IUPAC Name:
1-(bromomethyl)-3-(difluoromethoxy)-5-fluorobenzene
Sinonimi:
1-(bromomethyl)-3-(difluoromethoxy)-5-fluorobenzene
Peso molecolare (g/mol):
255.03
Formula molecolare:
C8H6BrF3O
InChi Key:
SZFFWIBFEJPELW-UHFFFAOYSA-N
SMILES:
FC(F)OC1=CC(F)=CC(CBr)=C1
1
–
1
di
1
risultati
1
5-(Difluoromethoxy)-3-fluorobenzyl bromide, JRD, 97%, Thermo Scientific™
CAS: 1017779-39-3 Formula molecolare: C8H6BrF3O Molecular Weight (g/mol): 255.034 Numero MDL: MFCD09832362 InChI Key: SZFFWIBFEJPELW-UHFFFAOYSA-N Sinonimo: 1-bromomethyl-3-difluoromethoxy-5-fluorobenzene,5-difluoromethoxy-3-fluorobenzyl bromide, jrd,5-difluoromethoxy-3-fluorobenzyl bromide,5-bromomethyl-3-fluorophenoxy difluoromethane PubChem CID: 46737678 IUPAC Name: 1-(bromomethyl)-3-(difluoromethoxy)-5-fluorobenzene SMILES: C1=C(C=C(C=C1OC(F)F)F)CBr