CAS RN 102-39-6
CAS RN 102-39-6
IUPAC Name:
2-[3-(carboxymethoxy)phenoxy]acetic acid
Sinonimi:
3-(carboxymethoxy)phenoxyacetic acid
Peso molecolare (g/mol):
226.18
Formula molecolare:
C10H10O6
InChi Key:
ZVMAGJJPTALGQB-UHFFFAOYSA-N
SMILES:
OC(=O)COC1=CC(OCC(O)=O)=CC=C1
1
–
1
di
1
risultati
1
Resorcinol-O,O'-diacetic acid, 97+%, Thermo Scientific™
CAS: 102-39-6 Formula molecolare: C10H10O6 Molecular Weight (g/mol): 226.184 Numero MDL: MFCD00016696 InChI Key: ZVMAGJJPTALGQB-UHFFFAOYSA-N Sinonimo: resorcinol-o,o'-diacetic acid,m-phenylenedioxydi acetic acid,2-3-carboxymethoxy phenoxy acetic acid,2,2'-1,3-phenylenebis oxy diacetic acid,2,2'-1,3-phenylenebis oxy bis-acetic acid,acetic acid,2,2'-1,3-phenylenebis oxy bis,3-carboxymethoxy phenoxyacetic acid,m-phenylenedioxy diacetic acid,m-phenylenedioxydi PubChem CID: 66884 IUPAC Name: 2-[3-(carboxymethoxy)phenoxy]acetic acid SMILES: C1=CC(=CC(=C1)OCC(=O)O)OCC(=O)O