CAS RN 102065-89-4
CAS RN 102065-89-4
IUPAC Name:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
3-Aminoazetidine dihydrochloride, 95%, Thermo Scientific Chemicals
CAS: 102065-89-4 Formula molecolare: C3H8N2 Molecular Weight (g/mol): 72.111 Numero MDL: MFCD09910173 InChI Key: FDPKMJDUXJFKOI-UHFFFAOYSA-N Sinonimo: 3-aminoazetidine,3-azetidinamine,acmc-209y1n,3-aminoazetidine dihy drochloric PubChem CID: 1516506 IUPAC Name: azetidin-3-amine SMILES: C1C(CN1)N