CAS RN 102851-06-9
CAS RN 102851-06-9
IUPAC Name:
cyano(3-phenoxyphenyl)methyl (2R)-2-{[2-chloro-4-(trifluoromethyl)phenyl]amino}-3-methylbutanoate
Sinonimi:
tau-fluvalinate
Peso molecolare (g/mol):
502.92
Formula molecolare:
C26H22ClF3N2O3
InChi Key:
INISTDXBRIBGOC-XMMISQBUSA-N
SMILES:
CC(C)[C@@H](NC1=CC=C(C=C1Cl)C(F)(F)F)C(=O)OC(C#N)C1=CC=CC(OC2=CC=CC=C2)=C1