CAS RN 10294-50-5
CAS RN 10294-50-5
IUPAC Name:
tris(λ²-cobalt(2+)) octahydrate diphosphate
Sinonimi:
tris(λ²-cobalt(2+)) octahydrate diphosphate
Peso molecolare (g/mol):
510.86
Formula molecolare:
Co3H16O16P2
InChi Key:
QOQSIXJUYVUEMP-UHFFFAOYSA-H
SMILES:
O.O.O.O.O.O.O.O.[Co++].[Co++].[Co++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
1
–
1
di
1
risultati
1
Cobalt(II) phosphate octahydrate, Thermo Scientific Chemicals
CAS: 10294-50-5 Formula molecolare: Co3H16O16P2 Molecular Weight (g/mol): 510.859 Numero MDL: MFCD00016028 InChI Key: QOQSIXJUYVUEMP-UHFFFAOYSA-H Sinonimo: cobalt ii phosphate octahydrate,cobaltous phosphate octahydrate,cobalt phosphate octahydrate,phosphoric acid, cobalt 2+ salt 2:3 , octahydrate,acmc-20ale3,3co.2po4.8h2o,cobalt 2+ diphosphate octahydrate PubChem CID: 202398 IUPAC Name: cobalt(2+);diphosphate;octahydrate SMILES: O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]