CAS RN 103057-44-9
CAS RN 103057-44-9
IUPAC Name:
tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate
Sinonimi:
tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate
Peso molecolare (g/mol):
187.24
Formula molecolare:
C9H17NO3
InChi Key:
APCBTRDHCDOPNY-ZETCQYMHSA-N
SMILES:
CC(C)(C)OC(=O)N1CC[C@H](O)C1
1
–
1
di
1
risultati
1
(+/-)-1-Boc-3-hydroxypyrrolidine, 97%, Thermo Scientific Chemicals
CAS: 103057-44-9 Formula molecolare: C9H17NO3 Molecular Weight (g/mol): 187.239 Numero MDL: MFCD04038535 InChI Key: APCBTRDHCDOPNY-UHFFFAOYSA-N Sinonimo: 1-boc-3-hydroxypyrrolidine,1-boc-3-pyrrolidinol,n-boc-3-hydroxypyrrolidine,n-boc-3-pyrrolidinol,1-n-boc-3-hydroxypyrrolidine,1-pyrrolidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester,1-tert-butoxycarbonyl-3-pyrrolidinol,3-hydroxy-pyrrolidine-1-carboxylic acid tert-butyl ester,tert-butyl 3-hydroxypyrrolidinecarboxylate,1-tert-butoxycarbonyl-3-hydroxypyrrolidine PubChem CID: 4416939 IUPAC Name: tert-butyl 3-hydroxypyrrolidine-1-carboxylate SMILES: CC(C)(C)OC(=O)N1CCC(C1)O