CAS RN 10342-83-3
CAS RN 10342-83-3
IUPAC Name:
1-(4-bromophenyl)propan-1-one
Sinonimi:
1-(4-bromophenyl)propan-1-one
Peso molecolare (g/mol):
213.07
Formula molecolare:
C9H9BrO
InChi Key:
UOMOSYFPKGQIKI-UHFFFAOYSA-N
SMILES:
CCC(=O)C1=CC=C(Br)C=C1
1
–
1
di
1
risultati
1
4'-Bromopropiophenone, 97%, Thermo Scientific Chemicals
CAS: 10342-83-3 Formula molecolare: C9H9BrO Molecular Weight (g/mol): 213.07 Numero MDL: MFCD00000106 InChI Key: UOMOSYFPKGQIKI-UHFFFAOYSA-N Sinonimo: 4'-bromopropiophenone,1-4-bromophenyl propan-1-one,p-bromopropiophenone,4-bromopropiophenone,propiophenone, 4'-bromo,4-bromophenyl ethyl ketone,1-propanone, 1-4-bromophenyl,1-4-bromophenyl-1-propanone,aurora 22532 PubChem CID: 66312 IUPAC Name: 1-(4-bromophenyl)propan-1-one SMILES: CCC(=O)C1=CC=C(Br)C=C1