CAS RN 1034566-05-6
CAS RN 1034566-05-6
IUPAC Name:
2-bromo-1-phenyl-1H-imidazole
Sinonimi:
2-bromo-1-phenylimidazole
Peso molecolare (g/mol):
223.07
Formula molecolare:
C9H7BrN2
InChi Key:
KKPQUNLAAJJUCR-UHFFFAOYSA-N
SMILES:
BrC1=NC=CN1C1=CC=CC=C1
1
–
1
di
1
risultati
1
2-bromo-1-phenyl-1H-imidazole, Thermo Scientific™
CAS: 1034566-05-6 Formula molecolare: C9H7BrN2 Molecular Weight (g/mol): 223.07 InChI Key: KKPQUNLAAJJUCR-UHFFFAOYSA-N Sinonimo: 2-bromo-1-phenyl-1h-imidazole,2-bromo-1-phenylimidazole PubChem CID: 45594301