CAS RN 103573-92-8
CAS RN 103573-92-8
IUPAC Name:
{4-[(1H-imidazol-1-yl)methyl]phenyl}methanol
Sinonimi:
[4-(imidazol-1-ylmethyl)phenyl]methanol
Peso molecolare (g/mol):
188.23
Formula molecolare:
C11H12N2O
InChi Key:
FSLVBXCLXDGFRE-UHFFFAOYSA-N
SMILES:
OCC1=CC=C(CN2C=CN=C2)C=C1
1
–
1
di
1
risultati
1
[4-(1H-Imidazol-1-ylmethyl)phenyl]methanol, ≥97%, Thermo Scientific™
CAS: 103573-92-8 Formula molecolare: C11H12N2O Molecular Weight (g/mol): 188.23 Numero MDL: MFCD08271921 InChI Key: FSLVBXCLXDGFRE-UHFFFAOYSA-N Sinonimo: 4-1h-imidazol-1-ylmethyl phenyl methanol,4-imidazol-1-ylmethyl phenyl methanol,4-1h-imidazol-1-yl methyl phenyl methanol,d0n8qd,4-imidazolylmethyl phenyl methan-1-ol,4-1h-imidazole-1-ylmethyl benzenemethanol,4-1h-imidazol-1-yl methyl phenylmethanol,benzenemethanol,4-1h-imidazol-1-ylmethyl PubChem CID: 15594569 IUPAC Name: [4-(imidazol-1-ylmethyl)phenyl]methanol SMILES: C1=CC(=CC=C1CN2C=CN=C2)CO