CAS RN 104-98-3
CAS RN 104-98-3
IUPAC Name:
(2E)-3-(1H-imidazol-5-yl)prop-2-enoic acid
Sinonimi:
urocanic acid
Peso molecolare (g/mol):
138.13
Formula molecolare:
C6H6N2O2
InChi Key:
LOIYMIARKYCTBW-OWOJBTEDSA-N
SMILES:
OC(=O)\C=C\C1=CN=CN1
1
–
1
di
1
risultati
1
Urocanic acid, 98%, Thermo Scientific Chemicals
CAS: 104-98-3 Formula molecolare: C6H6N2O2 Molecular Weight (g/mol): 138.13 Numero MDL: MFCD00005203 InChI Key: LOIYMIARKYCTBW-UPHRSURJSA-N Sinonimo: urocanic acid,4-imidazoleacrylic acid,trans-urocanic acid,urocaninic acid,imidazoleacrylic acid,5-imidazoleacrylic acid,urocanate,imidazole-4-acrylic acid,2e-3-1h-imidazol-4-yl acrylic acid,3-1h-imidazol-4-yl-2-propenoic acid PubChem CID: 1549103 ChEBI: CHEBI:30818 IUPAC Name: (Z)-3-(1H-imidazol-5-yl)prop-2-enoic acid SMILES: C1=C(NC=N1)C=CC(=O)O