CAS RN 104113-71-5
CAS RN 104113-71-5
IUPAC Name:
1-(naphthalen-1-yl)piperazine hydrochloride
Sinonimi:
naphthylpiperazine hydrochloride
Peso molecolare (g/mol):
248.75
Formula molecolare:
C14H17ClN2
InChi Key:
ZYVYPNZFOCZLEM-UHFFFAOYSA-N
SMILES:
Cl.C1CN(CCN1)C1=C2C=CC=CC2=CC=C1
1
–
1
di
1
risultati
1
1-(1-Naphthyl)piperazine hydrochloride, Thermo Scientific Chemicals
CAS: 104113-71-5 Formula molecolare: C14H17ClN2 Molecular Weight (g/mol): 248.754 Numero MDL: MFCD00055161 InChI Key: ZYVYPNZFOCZLEM-UHFFFAOYSA-N Sinonimo: 1-1-naphthyl piperazine hydrochloride,1-naphthalen-1-yl piperazine hydrochloride,naphthylpiperazine hydrochloride,1-naphthalen-1-ylpiperazine hydrochloride,1-naphthalen-1-yl-piperazine hydrochloride,piperazine,1-1-naphthalenyl,zlchem 1062,1-1-naphthyl piperazine hcl,1-naphthalen-1-yl-piperazine hcl salt,n-1-naphthyl piperazine hydrochloride PubChem CID: 2737391 IUPAC Name: 1-naphthalen-1-ylpiperazine;hydrochloride SMILES: C1CN(CCN1)C2=CC=CC3=CC=CC=C32.Cl