CAS RN 104632-25-9
CAS RN 104632-25-9
IUPAC Name:
dihydrogen (6S)-N6-propyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine dichloride
Sinonimi:
dihydrogen pramipexole dichloride
Peso molecolare (g/mol):
284.24
Formula molecolare:
C10H19Cl2N3S
InChi Key:
QMNWXHSYPXQFSK-KLXURFKVSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].CCCN[C@H]1CCC2=C(C1)SC(N)=N2
1
–
1
di
1
risultati
1
Pramipexole dihydrochloride, Tocris Bioscience™
CAS: 104632-25-9 Formula molecolare: C10H19Cl2N3S Molecular Weight (g/mol): 284.243 InChI Key: QMNWXHSYPXQFSK-KLXURFKVSA-N Sinonimo: pramipexole dihydrochloride,mirapex,pramipexole hydrochloride,s-n6-propyl-4,5,6,7-tetrahydrobenzo d thiazole-2,6-diamine dihydrochloride,mirapex er,unii-4r2hd0m28n,s-pramipexole dihydrochloride,bi-sifrol PubChem CID: 119569 ChEBI: CHEBI:51148 IUPAC Name: (6S)-6-N-propyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine;dihydrochloride SMILES: CCCNC1CCC2=C(C1)SC(=N2)N.Cl.Cl