CAS RN 10465-81-3
CAS RN 10465-81-3
IUPAC Name:
N-[(piperidine-1-carbonyl)imino]piperidine-1-carboxamide
Sinonimi:
N-(piperidine-1-carbonylimino)piperidine-1-carboxamide
Peso molecolare (g/mol):
252.32
Formula molecolare:
C12H20N4O2
InChi Key:
OQJBFFCUFALWQL-UHFFFAOYSA-N
SMILES:
O=C(N=NC(=O)N1CCCCC1)N1CCCCC1
1
–
1
di
1
risultati
1
1,1'-(Azodicarbonyl)dipiperidine, 97%, Thermo Scientific Chemicals
CAS: 10465-81-3 Formula molecolare: C12H20N4O2 Molecular Weight (g/mol): 252.32 Numero MDL: MFCD00010111 InChI Key: OQJBFFCUFALWQL-UHFFFAOYSA-N Sinonimo: 1,1'-azodicarbonyl-dipiperidine,1,1-azodicarbonyl dipiperidine,1,1'-azodicarbonyl dipiperidine,addp,diazene-1,2-diylbis piperidin-1-ylmethanone,azodicarboxylic acid dipiperidine,ne-n-piperidine-1-carbonylimino piperidine-1-carboxamide,1,1'-azodicarbonyl dipiperidine addp,piperidyl piperidylcarbonyl diazenyl ketone,e-diazene-1,2-diylbis piperidin-1-ylmethanone PubChem CID: 5702657 IUPAC Name: (NE)-N-(piperidine-1-carbonylimino)piperidine-1-carboxamide SMILES: O=C(N=NC(=O)N1CCCCC1)N1CCCCC1