CAS RN 10476-81-0
CAS RN 10476-81-0
IUPAC Name:
strontium(2+) dibromide
Sinonimi:
strontium bromide
Peso molecolare (g/mol):
247.43
Formula molecolare:
Br2Sr
InChi Key:
YJPVTCSBVRMESK-UHFFFAOYSA-L
SMILES:
[Br-].[Br-].[Sr++]
1
–
2
di
2
risultati
1
Strontium bromide, anhydrous, 99% (metals basis excluding Ba), Ba 0.1% max, Thermo Scientific Chemicals
CAS: 10476-81-0 Formula molecolare: Br2Sr Molecular Weight (g/mol): 247.428 Numero MDL: MFCD00049548 InChI Key: YJPVTCSBVRMESK-UHFFFAOYSA-L Sinonimo: acmc-20ajmt,strontium bromide srbr2,strontium 2+ ion dibromide PubChem CID: 5360351 IUPAC Name: strontium;dibromide SMILES: [Br-].[Br-].[Sr+2]
2
Strontium bromide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals
CAS: 10476-81-0 Formula molecolare: Br2Sr Molecular Weight (g/mol): 247.428 Numero MDL: MFCD00049548 InChI Key: YJPVTCSBVRMESK-UHFFFAOYSA-L Sinonimo: acmc-20ajmt,strontium bromide srbr2,strontium 2+ ion dibromide PubChem CID: 5360351 IUPAC Name: strontium;dibromide SMILES: [Br-].[Br-].[Sr+2]