CAS RN 10476-86-5
CAS RN 10476-86-5
IUPAC Name:
strontium(2+) diiodide
Sinonimi:
strontium iodide
Peso molecolare (g/mol):
341.43
Formula molecolare:
I2Sr
InChi Key:
KRIJWFBRWPCESA-UHFFFAOYSA-L
SMILES:
[Sr++].[I-].[I-]
1
–
2
di
2
risultati
1
Strontium iodide, anhydrous, 99% (metals basis), Thermo Scientific Chemicals
CAS: 10476-86-5 Numero MDL: MFCD00049557
2
Strontium iodide, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals
CAS: 10476-86-5 Formula molecolare: SrI2 Numero MDL: MFCD00049557