CAS RN 1047660-72-9
CAS RN 1047660-72-9
IUPAC Name:
triethyl(hept-1-yn-1-yl)silane
Sinonimi:
triethyl(hept-1-yn-1-yl)silane
Peso molecolare (g/mol):
210.44
Formula molecolare:
C13H26Si
InChi Key:
KIKRQMVMQSYDCD-UHFFFAOYSA-N
SMILES:
CCCCCC#C[Si](CC)(CC)CC
1
–
1
di
1
risultati
1
1-Triethylsilyl-1-heptyne, 95%, Thermo Scientific™
CAS: 1047660-72-9 Formula molecolare: C13H26Si Molecular Weight (g/mol): 210.436 Numero MDL: MFCD18800797 InChI Key: KIKRQMVMQSYDCD-UHFFFAOYSA-N Sinonimo: 1-triethylsilyl-1-heptyne,triethyl hept-1-yn-1-yl silane PubChem CID: 99738219 IUPAC Name: triethyl(hept-1-ynyl)silane SMILES: CCCCCC#C[Si](CC)(CC)CC