CAS RN 104784-51-2
CAS RN 104784-51-2
IUPAC Name:
trimethyl[2-(naphthalen-1-yl)ethynyl]silane
Sinonimi:
trimethyl[2-(naphthalen-1-yl)ethynyl]silane
Peso molecolare (g/mol):
224.38
Formula molecolare:
C15H16Si
InChi Key:
WATBCTJRCLNXNF-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)C#CC1=C2C=CC=CC2=CC=C1
1
–
1
di
1
risultati
1
1-(1-Naphthyl)-2-(trimethylsilyl)acetylene, 97%, Thermo Scientific Chemicals
CAS: 104784-51-2 Formula molecolare: C15H16Si Molecular Weight (g/mol): 224.378 Numero MDL: MFCD04039886 InChI Key: WATBCTJRCLNXNF-UHFFFAOYSA-N Sinonimo: 1-1-naphthyl-2-trimethylsilyl acetylene,trimethyl 2-naphthalen-1-ylethynyl silane,trimethyl 2-naphthalen-1-yl ethynyl silane,1-naphthylethynyl trimethylsilane,1-trimethylsilylethynyl naphthalene,1-trimethylsilyl ethynyl naphthalene,trimethyl naphthalen-1-ylethynyl silane,trimethyl naphthalen-1-yl ethynyl silane,trimethylnaphthalen-1-ylethynylsilane PubChem CID: 4438232 IUPAC Name: trimethyl(2-naphthalen-1-ylethynyl)silane SMILES: C[Si](C)(C)C#CC1=CC=CC2=CC=CC=C21