CAS RN 10489-74-4
CAS RN 10489-74-4
IUPAC Name:
ethyl (N'-hydroxycarbamimidoyl)formate
Sinonimi:
ethyl (N'-hydroxycarbamimidoyl)formate
Peso molecolare (g/mol):
132.12
Formula molecolare:
C4H8N2O3
InChi Key:
QGYKRMZPOOILBA-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(N)=NO
1
–
1
di
1
risultati
1
Ethyl 2-oximinooxamate, 98%, Thermo Scientific Chemicals
CAS: 10489-74-4 Formula molecolare: C4H8N2O3 Molecular Weight (g/mol): 132.12 Numero MDL: MFCD03013442 MFCD14666500 MFCD03013442 InChI Key: QGYKRMZPOOILBA-UHFFFAOYSA-N Sinonimo: ethyl 2-oximinooxamate,ethyl 2-amino-2-hydroxyimino acetate,ethyl z-2-amino-2-hydroxyimino acetate,ethyl 2-hydroxyamino-2-iminoacetate,ethyl n'-hydroxycarbamimidoyl formate,ethyl aminohydroxyiminoacetate,acetic acid, 2-amino-2-hydroxyimino-, ethyl ester, 2z,ethyl amino hydroxyimino ethanoate,ethyl n-hydroxycarbamimidoyl formate,amino-hydroxyimino-acetic acid ethyl ester PubChem CID: 9614369 IUPAC Name: ethyl (2Z)-2-amino-2-hydroxyiminoacetate SMILES: CCOC(=O)C(N)=NO