CAS RN 104992-55-4
CAS RN 104992-55-4
IUPAC Name:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
Thermo Scientific™ TheraPure™ Bz-rA Phosphoramidite, 20-400 finish bottle
Bz-rA CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with Benzoyl (Bz) base protection, for use in oligonucleotide synthesis.
Promozioni disponibili
2
Thermo Scientific™ TheraPure™ Bz-rA Phosphoramidite, serum vial bottle
Bz-rA CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with Benzoyl (Bz) base protection, for use in oligonucleotide synthesis.
3
Thermo Scientific™ TheraPure™ Bz-rA Phosphoramidite, wide mouth bottle
Bz-rA CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with Benzoyl (Bz) base protection, for use in oligonucleotide synthesis.