CAS RN 105-11-3
CAS RN 105-11-3
IUPAC Name:
N-(4-nitrosophenyl)hydroxylamine
Sinonimi:
N-(4-nitrosophenyl)hydroxylamine
Peso molecolare (g/mol):
138.13
Formula molecolare:
C6H6N2O2
InChi Key:
DZCCLNYLUGNUKQ-UHFFFAOYSA-N
SMILES:
ONC1=CC=C(C=C1)N=O
1
–
2
di
2
risultati
1
p-Benzoquinone dioxime, 95%, Thermo Scientific Chemicals
CAS: 105-11-3 Formula molecolare: C6H6N2O2 Molecular Weight (g/mol): 138.13 Numero MDL: MFCD00063636 InChI Key: LNHURPJLTHSVMU-CGXWXWIYSA-N Sinonimo: 1,4-benzoquinone dioxime,p-benzoquinonedioxime,p-benzoquinone dioxime,p-quinone dioxime,quinone dioxime,benzoquinone dioxime,p-quinone oxime,actor q,dioxime p-benzoquinone,dibenzo pqd PubChem CID: 7737 IUPAC Name: N-(4-nitrosophenyl)hydroxylamine SMILES: ON=C1C=CC(C=C1)=NO