CAS RN 105250-17-7
CAS RN 105250-17-7
IUPAC Name:
2-amino-4-(hydroxymethyl)pyridin-1-ium
Sinonimi:
2-amino-4-(hydroxymethyl)pyridin-1-ium
Peso molecolare (g/mol):
125.15
Formula molecolare:
C6H9N2O
InChi Key:
ZRJJXXDQIQFZBW-UHFFFAOYSA-O
SMILES:
NC1=CC(CO)=CC=[NH+]1
1
–
2
di
2
risultati
1
(2-Aminopyrid-4-yl)methanol, 97%, Thermo Scientific Chemicals
CAS: 105250-17-7 Formula molecolare: C6H9N2O Molecular Weight (g/mol): 125.15 Numero MDL: MFCD03791261 InChI Key: ZRJJXXDQIQFZBW-UHFFFAOYSA-O Sinonimo: 2-aminopyridin-4-yl methanol,2-amino-pyridin-4-yl-methanol,2-aminopyridine-4-methanol,2-amino-4-pyridinylmethanol,4-pyridinemethanol, 2-amino,2-aminopyridin-4-yl-methanol,2-amino-4-pyridinemethanol,2-amino-4-pyridinyl methanol,2-amino-4-hydroxymethylpyridine,2-amino-4-pyridyl methan-1-ol PubChem CID: 1515296 IUPAC Name: (2-aminopyridin-4-yl)methanol SMILES: NC1=CC(CO)=CC=[NH+]1