CAS RN 10538-48-4
CAS RN 10538-48-4
IUPAC Name:
3-(2,3-dimethoxyphenyl)propanoate
Sinonimi:
3-(2,3-dimethoxyphenyl)propanoate
Peso molecolare (g/mol):
209.22
Formula molecolare:
C11H13O4
InChi Key:
QPHAUUNQQLRYCJ-UHFFFAOYSA-M
SMILES:
COC1=CC=CC(CCC([O-])=O)=C1OC
1
–
1
di
1
risultati
1
3-(2,3-Dimethoxyphenyl)propionic acid, 96%, Thermo Scientific™
CAS: 10538-48-4 Formula molecolare: C11H14O4 Molecular Weight (g/mol): 210.229 Numero MDL: MFCD00029821 InChI Key: QPHAUUNQQLRYCJ-UHFFFAOYSA-N PubChem CID: 314929 IUPAC Name: 3-(2,3-dimethoxyphenyl)propanoic acid SMILES: COC1=CC=CC(=C1OC)CCC(=O)O