CAS RN 10541-83-0
CAS RN 10541-83-0
IUPAC Name:
4-(methylamino)benzoic acid
Sinonimi:
N-methyl-4-aminobenzoate
Peso molecolare (g/mol):
151.17
Formula molecolare:
C8H9NO2
InChi Key:
ZVIDMSBTYRSMAR-UHFFFAOYSA-N
SMILES:
CNC1=CC=C(C=C1)C(O)=O
1
–
1
di
1
risultati
1
4-(Methylamino)benzoic acid, 97%, Thermo Scientific Chemicals
CAS: 10541-83-0 Formula molecolare: C8H9NO2 Molecular Weight (g/mol): 151.165 Numero MDL: MFCD00002535 InChI Key: ZVIDMSBTYRSMAR-UHFFFAOYSA-N Sinonimo: 4-methylamino benzoic acid,benzoic acid, 4-methylamino,p-methylaminobenzoic acid,n-methyl-4-aminobenzoic acid,4-n-methylamino benzoic acid,n-methyl-4-aminobenzoate,benzoic acid, p-methylamino,4-methylamino-benzoic acid,p-methylamino benzoic acid,p-n-methylamino benzoic acid PubChem CID: 66345 ChEBI: CHEBI:7308 IUPAC Name: 4-(methylamino)benzoic acid SMILES: CNC1=CC=C(C=C1)C(=O)O