CAS RN 105628-64-6
CAS RN 105628-64-6
IUPAC Name:
tert-butyl N-(2-aminoethyl)-N-propylcarbamate
Sinonimi:
tert-butyl N-(2-aminoethyl)-N-propylcarbamate
Peso molecolare (g/mol):
202.3
Formula molecolare:
C10H22N2O2
InChi Key:
XQUOSTRRFPZQLE-UHFFFAOYSA-N
SMILES:
CCCN(CCN)C(=O)OC(C)(C)C
1
–
1
di
1
risultati
1
N-Boc-N-n-propylethylenediamine, 96%, Thermo Scientific™
CAS: 105628-64-6 Formula molecolare: C10H22N2O2 Molecular Weight (g/mol): 202.298 Numero MDL: MFCD06658368 InChI Key: XQUOSTRRFPZQLE-UHFFFAOYSA-N Sinonimo: tert-butyl n-2-aminoethyl-n-propylcarbamate,2-amino-ethyl-propyl-carbamic acid tert-butyl ester,n-boc-n-propylethylenediamine,tert-butyl 2-aminoethyl propyl carbamate,carbamic acid, 2-aminoethyl propyl-, 1,1-dimethylethyl ester 9ci,ambotzbnn1038,acmc-20m8mk,tert-butyl 2-aminoethyl propylcarbamate,tert-butyl n-2-azanylethyl-n-propyl-carbamate,n-2-aminoethyl-n-propylcarbamic acid tert-butyl ester PubChem CID: 14925148 IUPAC Name: tert-butyl N-(2-aminoethyl)-N-propylcarbamate SMILES: CCCN(CCN)C(=O)OC(C)(C)C