CAS RN 1392399-03-9
CAS RN 1392399-03-9
IUPAC Nome:
N-(1-benzylpiperidin-4-yl)-6,7-dimethoxy-2-(4-methyl-1,4-diazepan-1-yl)quinazolin-4-amine trihydrochloride
Sinonimi:
N-(1-benzylpiperidin-4-yl)-6,7-dimethoxy-2-(4-methyl-1,4-diazepan-1-yl)quinazolin-4-amine trihydrochloride
Peso molecolare (g/mol):
600.03
Formula molecolare:
C28H41Cl3N6O2
InChi Key:
FMURUEPQXKJIPS-UHFFFAOYSA-N
SMILES:
Cl.Cl.Cl.COC1=CC2=NC(=NC(NC3CCN(CC4=CC=CC=C4)CC3)=C2C=C1OC)N1CCCN(C)CC1
1
–
1
di
1
risultati
1
Tocris Bioscience™ BIX 01294
CAS: 1392399-03-9 Formula molecolare: C28H41Cl3N6O2