CAS RN 5888-33-5
CAS RN 5888-33-5
IUPAC Nome:
(1S,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl prop-2-enoate
Sinonimi:
isobornyl acrylate
Peso molecolare (g/mol):
208.3
Formula molecolare:
C13H20O2
InChi Key:
PSGCQDPCAWOCSH-BOURZNODSA-N
SMILES:
CC1(C)[C@H]2CC[C@]1(C)C(C2)OC(=O)C=C
1
–
1
di
1
risultati
1
Isobornyl acrylate, tech. 85%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
CAS: 5888-33-5 Formula molecolare: C13H20O2 Molecular Weight (g/mol): 208.30 Numero MDL: MFCD00080424 InChI Key: PSGCQDPCAWOCSH-BOURZNODSA-N Sinonimo: isobornyl acrylate,acrylic acid isobornyl ester,1,7,7-trimethyltricyclo 2.2.1 hepten-2-yl-2-propenoate PubChem CID: 122130333 IUPAC Name: [(3R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] prop-2-enoate SMILES: CC1(C)[C@H]2CC[C@]1(C)C(C2)OC(=O)C=C