CAS RN 70458-95-6
CAS RN 70458-95-6
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Pefloxacin mesylate, MedChemExpress
MedChemExpress Pefloxacin mesylate is a an antibacterial agent and prevents bacterial DNA replication by inhibiting DNA gyrase (topoisomerse) Target: DNA gyrase Pefloxacin is a synthetic chemotherapeutic agent used to treat severe and life-threatening bacterial infections.