CAS RN 7650-84-2
CAS RN 7650-84-2
IUPAC Nome:
diphenyl(propyl)phosphane
Sinonimi:
phosphine, diphenylpropyl-
Peso molecolare (g/mol):
228.28
Formula molecolare:
C15H17P
InChi Key:
AAXGWYDSLJUQLN-UHFFFAOYSA-N
SMILES:
CCCP(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
di
1
risultati
1
Diphenyl-n-propylphosphine, 97%
CAS: 7650-84-2 Formula molecolare: C15H17P Molecular Weight (g/mol): 228.28 Numero MDL: MFCD00015216 InChI Key: AAXGWYDSLJUQLN-UHFFFAOYSA-N Sinonimo: diphenyl-n-propylphosphine,phosphine, diphenylpropyl,propyldiphenylphosphine,n-propyldiphenylphosphine,diphenyl propyl phosphine,diphenylphosphinopropane,diphenyl propyl phosphane,diphenylphospinopropane,di phenyl-propylphosphane PubChem CID: 82100 IUPAC Name: diphenyl(propyl)phosphane SMILES: CCCP(C1=CC=CC=C1)C1=CC=CC=C1