Learn More
1,1-Diphenylethylene, 98%, Thermo Scientific Chemicals
1,1-Diphenylethylene, 98%, C14H12, CAS Number-530-48-3, unii-bx0l5b6lll, 1,1-diphenylethene, 1,1-diphenylethylene, 1-phenylethenyl benzene, 1-phenylvinyl benzene, ethene-1,1-diyldibenzene, bx0l5b6lll, .alpha.-phenylstyrene, benzene, 1,1'-ethenylidenebis, as-diphenylethylene, 250g, 1099062
50.20€ - 724.00€
Identificatori chimici
CAS | 530-48-3 |
---|---|
Formula molecolare | C14H12 |
Molecular Weight (g/mol) | 180.25 |
Numero MDL | MFCD00008583 |
InChI Key | ZMYIIHDQURVDRB-UHFFFAOYSA-N |
Sinonimo | 1,1-diphenylethylene, ethene-1,1-diyldibenzene, 1,1-diphenylethene, benzene, 1,1'-ethenylidenebis, 1-phenylethenyl benzene, unii-bx0l5b6lll, as-diphenylethylene, .alpha.-phenylstyrene, 1-phenylvinyl benzene, bx0l5b6lll |
PubChem CID | 10740 |
IUPAC Name | 1-phenylethenylbenzene |
SMILES | C=C(C1=CC=CC=C1)C2=CC=CC=C2 |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
11457077
|
Thermo Scientific Alfa Aesar
A14434.09 |
10 g |
50.20€
10g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11404250
|
Thermo Scientific Alfa Aesar
A14434.14 |
25 g |
100.00€
25g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11414250
|
Thermo Scientific Alfa Aesar
A14434.22 |
100 g |
303.00€
100g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11467077
|
Thermo Scientific Alfa Aesar
A14434.30 |
250 g |
724.00€
250g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
1,1-Diphenylethylene is used in the preparation of 2-chloro-1,1-diphenyl-ethene by reacting with benzeneseleninyl chloride and aluminum(III) chloride as reagents. It acts as an intermediate in organic synthesis as well as in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,1-Diphenylethylene is used in the preparation of 2-chloro-1,1-diphenyl-ethene by reacting with benzeneseleninyl chloride and aluminum(III) chloride as reagents. It acts as an intermediate in organic synthesis as well as in pharmaceuticals.
Solubility
Miscible with methanol, chloroform and ether. Insoluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents
Identificatori chimici
530-48-3 | |
180.25 | |
ZMYIIHDQURVDRB-UHFFFAOYSA-N | |
10740 | |
C=C(C1=CC=CC=C1)C2=CC=CC=C2 |
C14H12 | |
MFCD00008583 | |
1,1-diphenylethylene, ethene-1,1-diyldibenzene, 1,1-diphenylethene, benzene, 1,1'-ethenylidenebis, 1-phenylethenyl benzene, unii-bx0l5b6lll, as-diphenylethylene, .alpha.-phenylstyrene, 1-phenylvinyl benzene, bx0l5b6lll | |
1-phenylethenylbenzene |
Specifica
1,1-Diphenylethylene | |
5°C to 7°C | |
270°C to 271°C | |
C14H12 | |
MFCD00008583 | |
14,3323 | |
Miscible with methanol,chloroform and ether. Insoluble in water. | |
C=C(C1=CC=CC=C1)C2=CC=CC=C2 | |
180.25 | |
180.25 |
530-48-3 | |
1.023 | |
136°C (276°F) | |
1.6065 | |
1099062 | |
1,1-diphenylethylene, ethene-1,1-diyldibenzene, 1,1-diphenylethene, benzene, 1,1'-ethenylidenebis, 1-phenylethenyl benzene, unii-bx0l5b6lll, as-diphenylethylene, .alpha.-phenylstyrene, 1-phenylvinyl benzene, bx0l5b6lll | |
ZMYIIHDQURVDRB-UHFFFAOYSA-N | |
1-phenylethenylbenzene | |
10740 | |
98% |
Sicurezza e movimentazione
P261-P272-P280-P302+P352-P333+P313-P363-P501c
H317
missing translation for 'einecsNumber' : 208-482-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only