Learn More
1,2-Dimethoxy-4,5-dinitrobenzene, 96%, Thermo Scientific Chemicals
52.66€ - 178.89€
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
11352577
|
Thermo Scientific Alfa Aesar
L11706.03 |
1 g |
52.66€
1g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11362577
|
Thermo Scientific Alfa Aesar
L11706.06 |
5 g |
178.89€
5g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificatori chimici
3395-03-7 | |
MFCD00082695 |
Specifica
1,2-Dimethoxy-4,5-dinitrobenzene | |
131°C to 134°C | |
MFCD00082695 | |
2055812 | |
WFDHPWTYKOAFBJ-UHFFFAOYSA-N | |
1,2-dimethoxy-4,5-dinitrobenzene | |
222989 | |
96% |
3395-03-7 | |
C8H8N2O6 | |
UN2811 | |
4,5-dinitro veratrole, 1,2-dimethoxy-4,5-dinitro-benzene, 4,5-dimethoxy-1,2-dinitrobenzene, 4,5-dinitroveratrole, acmc-1ckho, ghl.pd_mitscher_leg0.933, benzene,1,2-dimethoxy-4,5-dinitro, benzene, 1,2-dimethoxy-4,5-dinitro | |
COC1=C(C=C(C(=C1)[N+](=O)[O-])[N+](=O)[O-])OC | |
228.16 | |
228.16 |
Sicurezza e movimentazione
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RUO – Research Use Only
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.