missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-(methylaminomethyl)azetidine, 95%, Thermo Scientific Chemicals

Codice prodotto. 15403678
Click to view available options
Quantità:
250 mg
500 mg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 1049730-81-5
Formula molecolare C10H20N2O2
Molecular Weight (g/mol) 200.28
Numero MDL MFCD08061965
InChI Key LNDGYTKISRFVOI-UHFFFAOYSA-N
Sinonimo tert-butyl 3-methylamino methyl azetidine-1-carboxylate, 1-boc-3-methylamino methyl azetidine, tert-butyl3-methylamino methyl azetidine-1-carboxylate, tert-butyl 3-methylamino methyl, 1-n-boc-3-methylaminomethylazetidine, 1-boc-3-methylaminomethyl azetidine, 1-n-boc-3-methylaminomethyl-azetidine, 3-methylamino methyl azetidine,n1-boc protected, 3-methylamino methyl azetidine, n1-boc protected
PubChem CID 46840011
IUPAC Name tert-butyl 3-(methylaminomethyl)azetidine-1-carboxylate
SMILES CNCC1CN(C1)C(=O)OC(C)(C)C

Specifica

Materiale o nome chimico 1-Boc-3-(methylaminomethyl)azetidine
Informazioni di solubilità Slightly soluble in water.
Peso formulazione 200.28
Percent Purity 95%
Quantità 250 mg

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'